Skip to Content
Merck
All Photos(1)

Key Documents

730092

Sigma-Aldrich

Trimethylamine-13C3,15N hydrochloride

99 atom % 13C, 98 atom % 15N

Synonym(s):

Trimethylammonium-13C3,15N chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(13CH3)315N HCl
Molecular Weight:
99.54
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % 15N

form

powder

mp

283-284 °C (dec) (lit.)

mass shift

M+4

SMILES string

Cl.[13CH3][15N]([13CH3])[13CH3]

InChI

1S/C3H9N.ClH/c1-4(2)3;/h1-3H3;1H/i1+1,2+1,3+1,4+1;

InChI key

SZYJELPVAFJOGJ-UJNKEPEOSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Marcin Ufnal et al.
The Canadian journal of cardiology, 30(12), 1700-1705 (2014-12-06)
Recent evidence suggests that an elevated plasma trimethylamine N-oxide (TMAO) level is associated with an increased risk of adverse cardiovascular events in humans; however, the mechanism is not clear. The aims of this study were to establish the plasma TMAO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service