715476
Pyrido[2,3-b]pyrazin-7-ylboronic acid pinacol ester
97%
Synonym(s):
7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrido[2,3-b]pyrazine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
Assay
97%
form
solid
mp
167-172 °C
storage temp.
2-8°C
SMILES string
CC1(C)OB(OC1(C)C)c2cnc3nccnc3c2
InChI
1S/C13H16BN3O2/c1-12(2)13(3,4)19-14(18-12)9-7-10-11(17-8-9)16-6-5-15-10/h5-8H,1-4H3
InChI key
QLYMAIYXJXENHJ-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service