Skip to Content
Merck
All Photos(1)

Key Documents

713732

Sigma-Aldrich

6-Amino-2-cyanobenzothiazole

97%

Synonym(s):

2-Cyano-6-aminobenzothiazole, 6-Amino-2-benzothiazolecarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5N3S
CAS Number:
Molecular Weight:
175.21
Beilstein:
1567902
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.5% (HPLC)
97%

form

crystals

functional group

nitrile

storage temp.

2-8°C

SMILES string

Nc1ccc2nc(sc2c1)C#N

InChI

1S/C8H5N3S/c9-4-8-11-6-2-1-5(10)3-7(6)12-8/h1-3H,10H2

InChI key

ZOHSEULTWOYIMS-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanyuan Yang et al.
Journal of biomedical nanotechnology, 14(8), 1396-1408 (2018-06-16)
Despite the great achievements of nanomedicines made in cancer chemotherapy, precise tumor targeting and deep penetration are still major challenges. Many nanomedicines can only passively accumulate near leaky site of tumor vessels through the enhanced permeability and retention (EPR) effect
Stefano Menegatti et al.
Advanced healthcare materials, 4(15), 2297-2305 (2015-09-16)
Combinations of polymer conjugates affording in situ gelation hold promise for treatment of pathological cavities (e.g., arthritis) and sustained drug release. In particular, hyaluronic acid (HA) functionalized with reactive groups is regarded as an excellent biomaterial due to its tunable
Pavlo V Khodakivskyi et al.
Science advances, 7(6) (2021-02-05)
The microbiome-produced enzyme bile salt hydrolase (BSH) plays a central role in human health, but its function remains unclear due to the lack of suitable methods for measuring its activity. Here, we have developed a novel optical tool based on
Nitin A Patil et al.
Bioconjugate chemistry, 30(3), 793-799 (2019-01-16)
Antisense oligonucleotide (ASO)-based drugs are emerging with great potential as therapeutic compounds for diseases with unmet medical needs. However, for ASOs to be effective as clinical entities, they should reach their intracellular RNA and DNA targets at pharmacologically relevant concentrations.
Jianfei Feng et al.
Communications biology, 3(1), 159-159 (2020-04-05)
Understanding protein folding under conditions similar to those found in vivo remains challenging. Folding occurs mainly vectorially as a polypeptide emerges from the ribosome or from a membrane translocon. Protein folding during membrane translocation is particularly difficult to study. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service