Skip to Content
Merck
All Photos(1)

Key Documents

712760

Sigma-Aldrich

6-Azido-6-deoxy-D-glucose

≥95% (HPLC)

Synonym(s):

6-Deoxy-6-azido-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11N3O5
CAS Number:
Molecular Weight:
205.17
Beilstein:
4906473
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95% (HPLC)

form

powder

optical activity

[α]/D +59.0±3.0°, c = 0.2 in H2O

composition

carbon, 34.4-35.8%
nitrogen, 20.0-21.0%

reaction suitability

reaction type: click chemistry

storage temp.

2-8°C

SMILES string

OC1O[C@H](CN=[N+]=[N-])[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C6H11N3O5/c7-9-8-1-2-3(10)4(11)5(12)6(13)14-2/h2-6,10-13H,1H2/t2-,3-,4+,5-,6?/m1/s1

InChI key

CMLRUUHRGSJVMD-GASJEMHNSA-N

Application

  • Visualization of Protein-Specific Glycation in Living Cells via Bioorthogonal Chemical Reporter.: This study by Shao et al. explores the use of 6-Azido-6-deoxy-ᴅ-glucose as a chemical reporter to visualize protein-specific glycation in living cells. The research demonstrates the application of bioorthogonal chemistry in detecting and imaging glycoconjugates, providing insights into cellular processes and potential pathways for drug delivery systems (Shao et al., 2022).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Georgieva et al.
Nucleic acids research, 48(15), e88-e88 (2020-07-28)
DNA synthesis is a fundamental requirement for cell proliferation and DNA repair, but no single method can identify the location, direction and speed of replication forks with high resolution. Mammalian cells have the ability to incorporate thymidine analogs along with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service