711217
5-Bromoisophthalic acid
Synonym(s):
5-Bromo-1,3-benzenedicarbonic acid, 5-Bromo-1,3-benzenedicarboxylic acid, 5-Bromoisophthalic acid
About This Item
Recommended Products
form
solid
mp
270-275 °C
functional group
bromo
carboxylic acid
SMILES string
OC(=O)c1cc(Br)cc(c1)C(O)=O
InChI
1S/C8H5BrO4/c9-6-2-4(7(10)11)1-5(3-6)8(12)13/h1-3H,(H,10,11)(H,12,13)
InChI key
JATKASGNRMGFSW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Preparation of manganese(II) bromoisophthalate bis(pyridyl)ethane coordination polymer
- Preparation of nickel(II) and cobalt(II) bromoisophthalate coordination polymers
- Preparation of copper bromoisophthalate bis(triazolyl)butane coordination polymer
- Preparation of cesium isophthalate crown ether complexes
- Hydrothermal preparation of zinc and cadmium bromoisophthalate polymeric complexes with dipyridyl-type coligands
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service