Skip to Content
Merck
All Photos(1)

Key Documents

700851

Sigma-Aldrich

(S)-Xyl-SDP

Synonym(s):

(S)-(−)-7,7′-Bis[di(3,5-dimethylphenyl)phosphino]-1,1′-spirobiindane, (S)-(−)-7,7′-Bis[di(3,5-dimethylphenyl)phosphino]-2,2′,3,3′-tetrahydro-1,1′-spirobiindene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H50P2
CAS Number:
Molecular Weight:
700.87
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

optical activity

[α]22/D −62.0°, c = 1 in chloroform

reaction suitability

reagent type: ligand

mp

>300 °C

functional group

phosphine

storage temp.

−20°C

SMILES string

Cc1cc(C)cc(c1)P(c2cc(C)cc(C)c2)c3cccc4CCC5(CCc6cccc(P(c7cc(C)cc(C)c7)c8cc(C)cc(C)c8)c56)c34

InChI

1S/C49H50P2/c1-31-19-32(2)24-41(23-31)50(42-25-33(3)20-34(4)26-42)45-13-9-11-39-15-17-49(47(39)45)18-16-40-12-10-14-46(48(40)49)51(43-27-35(5)21-36(6)28-43)44-29-37(7)22-38(8)30-44/h9-14,19-30H,15-18H2,1-8H3

InChI key

AZSBNBQMIMQOPG-UHFFFAOYSA-N

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service