Skip to Content
Merck
All Photos(1)

Key Documents

699594

Sigma-Aldrich

L-Thyroxine-(diphenyl-13C12)

99 atom % 13C, 97% (CP)

Synonym(s):

3,3′,5,5′′-Tetraiodo-L-thyronine-13C12, 3-[4-(4-Hydroxy-3,5-diiodophenoxy-13C6)-3,5-diiodophenyl-13C6]-L-alanine, T4-(ring-13C12)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO13C6H2(I)2O13C6H2(I)2CH2CH(NH2)CO2H
Molecular Weight:
788.78
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

223 °C (dec.) (lit.)

mass shift

M+12

storage temp.

2-8°C

SMILES string

N[C@@H](C[13c]1[13cH][13c](I)[13c](O[13c]2[13cH][13c](I)[13c](O)[13c](I)[13cH]2)[13c](I)[13cH]1)C(O)=O

InChI

1S/C15H11I4NO4/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23/h1-2,4-5,12,21H,3,20H2,(H,22,23)/t12-/m0/s1/i1+1,2+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,13+1,14+1

InChI key

XUIIKFGFIJCVMT-HTYRUWRGSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Other Notes

Bulk stock may be available for this product. Please contact the Stable Isotope Customer Service Group for pricing, availability and packaging at 937-859-1808 (1-800-448-9760 for US & Canada) or email us at [email protected]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Target Organs

Thyroid,Cardio-vascular system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service