695696
2-Hydroxybenzaldehyde phenylhydrazone
97%
Synonym(s):
Salicylic aldehyde phenylhydrazone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C13H12N2O
CAS Number:
Molecular Weight:
212.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
140-145 °C
SMILES string
Oc1ccccc1\C=N\Nc2ccccc2
InChI
1S/C13H12N2O/c16-13-9-5-4-6-11(13)10-14-15-12-7-2-1-3-8-12/h1-10,15-16H/b14-10+
InChI key
SERARPRVBWDEBA-GXDHUFHOSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service