Skip to Content
Merck
All Photos(4)

Key Documents

693871

Sigma-Aldrich

Calcium phosphate, amorphous

nanopowder, <150 nm particle size (BET)

Synonym(s):

ACP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca2O7P2 · H2O
CAS Number:
EC Number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanopowder

Quality Level

composition

Ca/P, ~1:1

surface area

>12 m2/g , typical

particle size

<150 nm (BET)

SMILES string

O.[Ca++].[Ca++].[O-]P([O-])(=O)OP([O-])([O-])=O

InChI

1S/2Ca.H4O7P2.H2O/c;;1-8(2,3)7-9(4,5)6;/h;;(H2,1,2,3)(H2,4,5,6);1H2/q2*+2;;/p-4

InChI key

JWETXQZDTWPZNK-UHFFFAOYSA-J

Legal Information

Product of Engi-Mat Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas P Diamantopoulos et al.
Journal of clinical rheumatology : practical reports on rheumatic & musculoskeletal diseases, 18(6), 310-311 (2012-07-27)
A 54-year-old woman with recurrent episodes of bilateral knee arthritis was admitted several times to the Department of Rheumatology. She was diagnosed to have chronic calcium pyrophosphate crystal arthritis. Interleukin 1β (IL-1β) plays a central role in the pathogenesis of
Christoph Robier et al.
Clinical chemistry and laboratory medicine, 50(2), 371-373 (2011-10-12)
Automated leukocyte differential counts of synovial fluid (SF) can be influenced by laboratory artefacts. Pseudoeosinophilia of SF has recently been first described in association with monosodium urate and calcium pyrophosphate crystals. This study compared automated measurements of the percentages of
Tim L Jansen et al.
Clinical and experimental rheumatology, 29(6), 1032-1039 (2011-12-03)
Many crystals may be found in arthritic joints. Rheumatologists are able to diagnose, with a high degree of probability, which crystals induce arthritis in the individual case. A definite diagnosis supported by polarisation microscopy may provide a firm basis for
Jae Hyup Lee et al.
Clinics in orthopedic surgery, 3(3), 238-244 (2011-09-13)
Porous β-calcium pyrophosphate (β-CPP) was developed to improve the fusion success of posterolateral lumbar fusion (PLF). The possibility of accomplishing PLF using a mixture of porous β-CPP and iliac bone was studied. This paper reports the radiologic results of PLF
Yan Shi
Protein & cell, 3(8), 564-570 (2012-06-22)
Phagocytosis and innate immune responses to solid structures are topics of interest and debate. Alum, monosodium urate, calcium pyrophosphate dehydrate, silica and by extension all solid entities draw varying degrees of attention from phagocytes, such as antigen presenting cells. For

Articles

Innovation in dental restorative materials is driven by the need for biocompatible and natural-appearing restoration alternatives. Conventional dental materials like amalgam and composite resins have inherent disadvantages.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service