Skip to Content
Merck
All Photos(1)

Key Documents

693006

Sigma-Aldrich

(S)-SEGPHOS®

≥94%

Synonym(s):

(S)-(−)-5,5′-Bis(diphenylphosphino)-4,4′-bi-1,3-benzodioxole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H28O4P2
CAS Number:
Molecular Weight:
610.57
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥94%

form

solid

optical activity

[α]20/D -11°, c = 0.5 in chloroform

mp

231-235 °C

functional group

phosphine

SMILES string

P(c8ccccc8)(c7ccccc7)c1c(c6c(cc1)OCO6)c2c3c(ccc2P(c5ccccc5)c4ccccc4)OCO3

InChI

1S/C38H28O4P2/c1-5-13-27(14-6-1)43(28-15-7-2-8-16-28)33-23-21-31-37(41-25-39-31)35(33)36-34(24-22-32-38(36)42-26-40-32)44(29-17-9-3-10-18-29)30-19-11-4-12-20-30/h1-24H,25-26H2

InChI key

RZZDRSHFIVOQAF-UHFFFAOYSA-N

Application

Chiral biaryl bisphosphine catalytic ligand used for:
  • Rhodium-catalyzed asymmetric formal olefination or cycloaddition of 1,3-dicarbonyl compounds with 1,6-diynes or 1,6-enynes
  • Stereoselective preparation of homoallylic alcohols via Ir-catalyzed stereoselective transfer hydrogenative crotylation of an allylic acetate with alcohols or aldehydes
  • Regio- and stereoselective preparation of axially chiral arylnaphthalene derivatives via rhodium-catalyzed [2+2+2] cycloaddition of diynes with naphthalenepropynoic acid derivatives
  • Diastereo- and enantioselective hydrogenation of α-amino-β-keto ester hydrochlorides catalyzed by an iridium complex
  • Preparation of dihydroisoquinolinones via nickel-catalyzed denitrogenative annulation of benzotriazinones with 1,3-dienes and alkenes
  • Stereoselective preparation of indanol derivatives via Rh-catalyzed enantioselective alkynylative cyclization of allenyl aldehydes with terminal alkynes

Legal Information

Sold in collaboration with Takasago for research purposes only. JP Registration No. 3148136
SEGPHOS is a registered trademark of Takasago Intl. Corp.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service