Skip to Content
Merck
All Photos(1)

Key Documents

691941

Sigma-Aldrich

3-Benzyladenine

95%

Synonym(s):

3-Benzyl-3H-purin-6-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N5
CAS Number:
Molecular Weight:
225.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

mp

268-275 °C

SMILES string

Nc1ncn(Cc2ccccc2)c3ncnc13

InChI

1S/C12H11N5/c13-11-10-12(15-7-14-10)17(8-16-11)6-9-4-2-1-3-5-9/h1-5,7-8H,6,13H2

InChI key

ZRRXJVVXTVXDII-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J Li et al.
Shi yan sheng wu xue bao, 27(4), 457-462 (1994-12-01)
The effect of purine on the meiosis of mouse oocytes was studied. When adenine and benzyladenine, a similar substance of adenine, was microinjected into the germinal vesicle of oocytes, the meiotic maturation of oocytes could be obviously arrested depending on
Yuki Ishii et al.
Biochimica et biophysica acta, 1643(1-3), 11-24 (2003-12-05)
Cytokinins are important purine derivatives that act as hormones to control many processes in plants. Cytokinins such as isopentenyladenine (IPA), kinetin and benzyladenine were very effective at inducing the granulocytic differentiation of human myeloid leukemia HL-60 cells. The metabolism of
V Prevost et al.
Carcinogenesis, 14(2), 199-204 (1993-02-01)
Immunoaffinity gels were prepared by coupling monoclonal antibody (Mab) EM-6-47 to protein A-Sepharose, and were used to make small columns retaining 3-alkyladenines (3-alkAde) of diverse structure. An analytical procedure for determination of 3-methyladenine (3-MeAde), 3-ethyladenine (3-EtAde), 3-(2-hydroxyethyl)adenine (3-HOEtAde) and 3-benzyladenine
H Yamada et al.
Plant & cell physiology, 42(9), 1017-1023 (2001-09-29)
Common histidine-to-aspartate (His-->Asp) phosphorelay is a paradigm of signal transduction in both prokaryotes and eukaryotes for the propagation of certain environmental stimuli, in which histidine (His)-kinases play central roles as sensors for environmental signals. For the higher plant, Arabidopsis thaliana

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service