68346
3-Methyl-1,5-pentanediol
≥98.0% (GC)
Synonym(s):
1,5-Dihydroxy-3-methylpentane, Diol MPD, MPD
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥98.0% (GC)
form
liquid
refractive index
n20/D 1.454
density
0.974 g/mL at 20 °C (lit.)
functional group
hydroxyl
SMILES string
CC(CCO)CCO
InChI
1S/C6H14O2/c1-6(2-4-7)3-5-8/h6-8H,2-5H2,1H3
InChI key
SXFJDZNJHVPHPH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
3-Methyl-1,5-pentanediol can be used as a reactant to synthesize:
- Enantioenrich saturated aza-heterocycles by reacting with various amines in the presence of iridium catalyst.
- Polyesters containing carbon-carbon double bonds via bis(nonafluorobutanesulfonyl)imide-catalyzed polycondensation with unsaturated cyclic anhydrides.
- α, ω-Dibromides by treating with aqueous HBr and an organic solvent.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
289.4 °F - closed cup
Flash Point(C)
143 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Frontiers in microbiology, 10, 3158-3158 (2020-02-11)
HigB-HigA is a bacterial toxin-antitoxin (TA) system in which the antitoxin HigA can mask the endoribonuclease activity of toxin HigB and repress the transcription of the TA operon by binding to its own promoter region. The opportunistic pathogen Pseudomonas aeruginosa
Molecules (Basel, Switzerland), 25(20) (2020-10-18)
The goal of this work was to develop a shape memory polymer (SMP) foam with visibility under both X-ray and magnetic resonance imaging (MRI) modalities. A porous polymeric material with these properties is desirable in medical device development for applications
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service