683159
Chiralyst P468
Umicore
Synonym(s):
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, Bis(1,5-cyclooctadiene)rhodium triflate
About This Item
Recommended Products
form
powder
reaction suitability
core: rhodium
reagent type: catalyst
mp
159 °C (dec. literature value) (lit.)
83-97 °C (dec. experimental value)
SMILES string
[Rh+].[O-]S(=O)(=O)C(F)(F)F.C1CC=CCCC=C1.C2CC=CCCC=C2
InChI
1S/2C8H12.CHF3O3S.Rh/c2*1-2-4-6-8-7-5-3-1;2-1(3,4)8(5,6)7;/h2*1-2,7-8H,3-6H2;(H,5,6,7);/q;;;+1/p-1/b2*2-1-,8-7-;;
InChI key
GOPBTCMAUMSOBX-QMDOQEJBSA-M
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.
Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.
Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.
Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service