679755
Hexafluorozirconic acid solution
50 wt. % in H2O
Synonym(s):
Dihydrogen hexafluorozirconate solution, Hydrogen zirconium fluoride
About This Item
Recommended Products
reaction suitability
core: zirconium
reagent type: catalyst
concentration
50 wt. % in H2O
density
1.512 g/mL at 25 °C
SMILES string
F.F.F[Zr](F)(F)F
InChI
1S/6FH.Zr/h6*1H;/q;;;;;;+4/p-4
InChI key
DXIGZHYPWYIZLM-UHFFFAOYSA-J
Looking for similar products? Visit Product Comparison Guide
Application
- Hexafluorozirconic acid based surface pretreatments on steel for corrosion resistance
- Preparation of titania photocatalyst synthesized from ionic-liquid-like precursor
- Synthesis of fluoride-releasing dental monomer
- As precursor to ZrO2 ceramic thin films
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service