Skip to Content
Merck
All Photos(1)

Key Documents

677426

Sigma-Aldrich

Iron-nickel alloy

nanopowder, <100 nm particle size (BET), ≥97%

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

description

Fe/Ni (0.55:0.45)

Quality Level

Assay

≥97%

form

nanopowder

composition

Fe, 55%
Ni, 45%

reaction suitability

reagent type: catalyst
core: nickel

surface area

>14.4 m2/g

particle size

<100 nm (BET)

Legal Information

Product of Engi-Mat Co.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Flam. Sol. 2 - Self-heat. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Electronic and Magnetic Structure of bcc Iron-Nickel Alloy
M. E. Elzain, A.A. Yousif, A.D. Al Rawas, A.M. Gismelseed
Journal of Physics. Condensed Matter : An Institute of Physics Journal, 11, 1545-1556 (1999)
L Dubrovinsky et al.
Physical review letters, 86(21), 4851-4854 (2001-06-01)
We have measured the pressure-volume (P-V) relations for cubic iron-nickel alloys for three different compositions: Fe 0.64Ni (0.36), Fe 0.55Ni (0.45), and Fe 0.20Ni (0.80). It is observed that for a certain pressure range the bulk modulus does not change

Articles

Mechanical alloying is a “brute force” method of affecting alloying and chemical reactions. The mixture of reactant powders and several balls are placed in the milling jar of a high-energy ball mill, for example, a shaker mill or a planetary mill.

Professor Hui Mao explores the use of superparamagnetic iron oxide nanoparticles (INOPs) that offer an alternate contrast-enhancing mechanism.

Prof. Yadong Yin discusses various synthesis methods of magnetite nanocrystals and their applications in different fields.

Prof. Yadong Yin discusses various synthesis methods of magnetite nanocrystals and their applications in different fields.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service