Skip to Content
Merck
All Photos(1)

Key Documents

674192

Sigma-Aldrich

Trioxo(triphenylsilyloxy)rhenium(VII)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15O4ReSi
CAS Number:
Molecular Weight:
509.60
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

core: rhenium
reagent type: catalyst

mp

108-110 °C

storage temp.

2-8°C

SMILES string

O=[Re](=O)(=O)O[Si](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C18H15OSi.3O.Re/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;;/h1-15H;;;;/q-1;;;;+1

InChI key

JFZHOPFMMQQPJF-UHFFFAOYSA-N

Application

Catalyst for:
  • Cycloisomerization reactions
  • Prins dimerization
  • Prins cyclization reactions

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service