671673
Methyl 4-(chlorocarbonyl)benzoate
≥95%
Synonym(s):
4-Chlorocarbonylbenzoic acid methyl ester, Methyl 4-(chloroformyl)benzoate, Methyl terephthaloyl chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H7ClO3
CAS Number:
Molecular Weight:
198.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥95%
≥95.0% (GC)
95.0-105.0% (AT)
form
solid
functional group
acyl chloride
ester
SMILES string
COC(=O)c1ccc(cc1)C(Cl)=O
InChI
1S/C9H7ClO3/c1-13-9(12)7-4-2-6(3-5-7)8(10)11/h2-5H,1H3
InChI key
CVXXHXPNTZBZEL-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service