Skip to Content
Merck

661414

Sigma-Aldrich

D-Glucose-6,6-d2

endotoxin tested, ≥98 atom % D, ≥99% (CP)

Synonym(s):

Deuterated Glucose, Labeled Glucose, Dextrose-6,6-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D2H10O6
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:


Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥98 atom % D

Assay

≥99% (CP)

form

powder

mp

150-152 °C (lit.)

suitability

endotoxin tested

mass shift

M+2

SMILES string

[2H]C([2H])(O)[C@H]1OC(O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i1D2

InChI key

WQZGKKKJIJFFOK-VARZTKRMSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
394238658847684457
assay

97%

assay

≥99.5%

assay

97% (HPLC)

assay

>99.9%

solubility

water, pH >9: soluble

solubility

water: miscible

solubility

-

solubility

chlorobenzene: soluble, organic solvents: soluble, toluene: soluble

mp

>300 °C

mp

-

mp

-

mp

-

Application

For R&D use only. Not for drug, household, or other uses. Any other use is the sole responsibility of the end user.

Endotoxin testing is done on the bulk form of the product before subdivision packaging. This bulk test does not guarantee that the subdivision or repackaged aliquot is endotoxin-free when it is received or used by the customer and does not imply suitability for any particular purpose. If the product must be endotoxin-free for the intended application, the product should be tested prior to actual use.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Qiang Wu et al.
Nucleic acids research, 43(9), 4614-4626 (2015-04-22)
Mutations in the RecQ DNA helicase gene BLM give rise to Bloom's syndrome, which is a rare autosomal recessive disorder characterized by genetic instability and cancer predisposition. BLM helicase is highly active in binding and unwinding G-quadruplexes (G4s), which are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service