Skip to Content
Merck
All Photos(1)

Key Documents

660221

Sigma-Aldrich

2-Bromo-2′,6′-diisopropoxy-1,1′-biphenyl

95%

Synonym(s):

2-Bromo-2′,6′-diisopropoxybiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21BrO2
CAS Number:
Molecular Weight:
349.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.562

bp

140 °C/0.2 mmHg

density

1.231 g/mL at 25 °C

functional group

bromo

SMILES string

CC(C)Oc1cccc(OC(C)C)c1-c2ccccc2Br

InChI

1S/C18H21BrO2/c1-12(2)20-16-10-7-11-17(21-13(3)4)18(16)14-8-5-6-9-15(14)19/h5-13H,1-4H3

InChI key

SHUUMGHTDLOJAU-UHFFFAOYSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service