Skip to Content
Merck
All Photos(2)

Key Documents

65290

Sigma-Aldrich

3-Methoxypropionitrile

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2CN
CAS Number:
Molecular Weight:
85.10
Beilstein:
1739284
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥98.0% (GC)

refractive index

n20/D 1.403 (lit.)
n20/D 1.403

bp

164-165 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

functional group

ether
nitrile

SMILES string

COCCC#N

InChI

1S/C4H7NO/c1-6-4-2-3-5/h2,4H2,1H3

InChI key

OOWFYDWAMOKVSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongting Wang et al.
Nanomaterials (Basel, Switzerland), 9(7) (2019-07-03)
A facile one-pot approach was developed for the synthesis of ZnO nanorods (NRs)/nanoparticles (NPs) architectures with controllable morphologies. The concrete state of existence of NPs and NRs could rationally be controlled through reaction temperature manipulation, i.e., reactions occured at 120
Liyou Lu et al.
Nanoscale research letters, 7(1), 293-293 (2012-06-08)
In this work, we report a direct synthesis of vertically aligned ZnO nanowires on fluorine-doped tin oxide-coated substrates using the chemical vapor deposition (CVD) method. ZnO nanowires with a length of more than 30 μm were synthesized, and dye-sensitized solar cells
Angel Ramon Hernández-Martínez et al.
International journal of molecular sciences, 14(2), 4081-4093 (2013-02-23)
Dye-Sensitized Solar Cells (DSSCs), based on TiO(2) and assembled using a dye from Beta vulgaris extract (BVE) with Tetraethylorthosilicate (TEOS), are reported. The dye BVE/TEOS increased its UV resistance, rendering an increase in the cell lifetime; the performance of these
Christopher Lausted et al.
Genome biology, 5(8), R58-R58 (2004-08-04)
DNA arrays are valuable tools in molecular biology laboratories. Their rapid acceptance was aided by the release of plans for a pin-spotting microarrayer by researchers at Stanford. Inkjet microarraying is a flexible, complementary technique that allows the synthesis of arrays
Yong Zhang et al.
ChemSusChem, 11(11), 1813-1823 (2018-05-10)
High-efficiency perovskite solar cells are generally fabricated by using highly pure (>99.99 %) PbI2 mixed with an organic iodide in polar aprotic solvents. However, the use of such an expensive chemical may impede progress toward large-scale industrial applications. Here, we report

Articles

Dye-sensitized solar cells directly convert sunlight to electricity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service