Skip to Content
Merck
All Photos(2)

Key Documents

640042

Sigma-Aldrich

4-Amino-1-Boc-piperidine

97%

Synonym(s):

1-Boc-4-piperidinamine, tert-Butyl 4-amino-1-piperidinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

SMILES string

CC(C)(C)OC(=O)N1CCC(N)CC1

InChI

1S/C10H20N2O2/c1-10(2,3)14-9(13)12-6-4-8(11)5-7-12/h8H,4-7,11H2,1-3H3

InChI key

LZRDHSFPLUWYAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Amino-1-Boc-piperidine can be used:
  • as a starting material in the synthesis of N-(3-(4-hydroxyphenyl)-propenoyl)-amino acid tryptamides, which can act as silent information regulator human type 2 (SIRT2) inhibitors[1]
  • to synthesize piperidine-substituted triazine derivatives[2]and piperidinylamino-diarylpyrimidine (pDAPY) derivatives as potent HIV-1 non-nucleoside reverse transcriptase inhibitors (HIV-1 NNRTIs)[3]
Employed in a microwave-assisted solid-phase synthesis of N-substituted piperidines via direct annulation of primary amines with resin-bound dimesylates.[4]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Novel piperidinylamino-diarylpyrimidine derivatives with dual structural conformations as potent HIV-1 non-nucleoside reverse transcriptase inhibitors"
Chen X, et al.
Bioorganic & Medicinal Chemistry, 23(24), 6593-6597 (2013)
"N-(3-(4-Hydroxyphenyl)-propenoyl)-amino acid tryptamides as SIRT2 inhibitors"
Kiviranta.HP, et al.
Bioorganic & Medicinal Chemistry, 17(09), 2448-2451 (2007)
Calum Macleod et al.
Journal of combinatorial chemistry, 8(1), 132-140 (2006-01-10)
The microwave-assisted solid-phase synthesis of piperazines, 3,9-diazaspiro[5.5]undecanes and 2,9-diazaspiro[5.5]undecanes is reported. The synthesis relies on the direct annulation of primary amines with resin-bound bismesylates. Critical to the success of this chemistry was the development of alpha-methyl benzyl carbamate resin linker.
"Synthesis and biological evaluation of piperidine-substituted triazine derivatives as HIV-1 non-nucleoside reverse transcriptase inhibitors"
Chen X, et al.
European Journal of Organic Chemistry, 51, 60- 66 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service