Skip to Content
Merck
All Photos(1)

Key Documents

633291

Sigma-Aldrich

N-Propionyloxy-d5-succinimide

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7D5H4NO4
CAS Number:
Molecular Weight:
176.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

mass shift

M+5

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C(=O)ON1C(=O)CCC1=O

InChI

1S/C7H9NO4/c1-2-7(11)12-8-5(9)3-4-6(8)10/h2-4H2,1H3/i1D3,2D2

InChI key

AASBXERNXVFUEJ-ZBJDZAJPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service