Skip to Content
Merck
All Photos(1)

Key Documents

616990

Sigma-Aldrich

Triphenylphosphine-d15

98 atom % D

Synonym(s):

Tri(phenyl-d5)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6D5)3P
CAS Number:
Molecular Weight:
277.38
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

bp

377 °C

mp

79-81 °C (lit.)

mass shift

M+15

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])P(c2c([2H])c([2H])c([2H])c([2H])c2[2H])c3c([2H])c([2H])c([2H])c([2H])c3[2H]

InChI

1S/C18H15P/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D,15D

InChI key

RIOQSEWOXXDEQQ-KLHTYYPYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

428.0 °F - closed cup

Flash Point(C)

220.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service