Skip to Content
Merck
All Photos(2)

Key Documents

613150

Sigma-Aldrich

3-(Trimethylsilyl)-1-propanesulfonic acid-d6 sodium salt

98 atom % D

Synonym(s):

2,2-Dimethyl-2-silapentane-5-sulfonate-d6 sodium salt, DSS-d6, Deuterated 3-(trimethylsilyl)-1-propanesulfonic acid sodium salt, Sodium 2-(trimethylsilyl)-1-propanesulfonate-d6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3Si(CD2)3SO3Na
CAS Number:
Molecular Weight:
224.36
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.11

isotopic purity

98 atom % D

Quality Level

form

solid

technique(s)

NMR: suitable
bio NMR: suitable

mp

118-120 °C (lit.)

mass shift

M+6

SMILES string

[Na+].[2H]C([2H])(C([2H])([2H])[Si](C)(C)C)C([2H])([2H])S([O-])(=O)=O

InChI

1S/C6H16O3SSi.Na/c1-11(2,3)6-4-5-10(7,8)9;/h4-6H2,1-3H3,(H,7,8,9);/q;+1/p-1/i4D2,5D2,6D2;

InChI key

HWEXKRHYVOGVDA-CHBZFOKHSA-M

Looking for similar products? Visit Product Comparison Guide

General description

3-(Trimethylsilyl)-1-propanesulfonic acid (also referred to as DSS (2,2-Dimethyl-2-silapentane-5-sulfonate)) is an internal standard for NMR (nuclear magnetic resonance) studies. It results in high intensity proton peaks which are easily identified. DSS also has negligible sensitivity towards pH and temperature changes.

Application

In exometabolome analysis using 1H-NMR (nuclear magnetic resonance) spectroscopy, 3-(Trimethylsilyl)-1-propanesulfonic acid-d6 sodium salt has been used as an internal standard for metabolite quantification. It has been used for the preparation of DMSO (dimethyl sulfoxide)/DSS solvent solution to make NMR leaf extract samples.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathleen A Stringer et al.
Shock (Augusta, Ga.), 44(3), 200-208 (2015-05-27)
Serum is a common sample of convenience for metabolomics studies. Its processing time can be lengthy and may result in the loss of metabolites including those of red blood cells (RBCs). Unlike serum, whole blood (WB) is quickly processed, minimizing
Metabolic signatures of GS-CHO cell clones associated with butyrate treatment and culture phase transition.
Carinhas N, et al.
Biotechnology and Bioengineering, 110, 3244-3257 (2013)
Niki Marie Zacharias et al.
Scientific reports, 7(1), 16159-16159 (2017-11-25)
The new oncologic paradigm of precision medicine is focused on identifying metabolic, proteomic, transcriptomic and genomic variabilities in tumors that can be exploited to tailor treatments and improve patient outcomes. Metabolic changes are a hallmark of cancer, and inhibition of
Mathias T Vangsoe et al.
Nutrients, 10(10) (2018-09-27)
Increased amino acid availability stimulates muscle protein synthesis (MPS), which is critical for maintaining or increasing muscle mass when combined with training. Previous research suggests that whey protein is superior to soy protein in regard to stimulating MPS and muscle
Francisco Madrid-Gambin et al.
Journal of agricultural and food chemistry, 66(27), 6997-7005 (2018-06-20)
High legume intake has been shown to have beneficial effects on the health of humans. The use of nutritional biomarkers, as a complement to self-reported questionnaires, could assist in evaluating dietary intake and downstream effects on human health. The aim

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service