Skip to Content
Merck
All Photos(1)

Key Documents

612022

Supelco

Nicotine solution

NMR reference standard, 2 mg/mL in chloroform-d (99.8 atom % D), TMS 0.05 %, NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2
Molecular Weight:
162.23
UNSPSC Code:
12142201
NACRES:
NA.24

grade

NMR reference standard
analytical standard

Quality Level

composition

TMS, 0.05%

concentration

2 mg/mL in chloroform-d (99.8 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

format

single component solution

Looking for similar products? Visit Product Comparison Guide

General description

NMR reference standard (2 mg/mL in chloroform-d (99.8 atom % D), TMS 0.05%) eliminates the sample preparation uncertainty when evaluating instrument performance and assures proper instrument performance in quantitative applications. These NMR reference standards allow the users to monitor key operational parameters such as pw90, sensitivity, resolution, and line shape.

Quantity

5 mm O.D. tube contains 0.700 mL.
Each pack contains 10 tubes.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service