Skip to Content
Merck
All Photos(1)

Key Documents

609099

Sigma-Aldrich

L-Threonine-15N

98 atom % 15N, 98% (CP)

Synonym(s):

(2S,3R)-2-Amino-3-hydroxybutyric acid-15N, 15N Labeled L-threonine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(15NH2)CO2H
Molecular Weight:
120.11
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

Quality Level

Assay

98% (CP)

form

solid

optical activity

[α]20/D −27.4°, c = 1 in H2O

technique(s)

bio NMR: suitable

mp

256 °C (dec.) (lit.)

mass shift

M+1

SMILES string

C[C@@H](O)[C@H]([15NH2])C(O)=O

InChI

1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3+/m1/s1/i5+1

InChI key

AYFVYJQAPQTCCC-SZEKAKMSSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service