Skip to Content
Merck
All Photos(1)

Key Documents

608661

Sigma-Aldrich

Nitrogen-14N2

99.99 atom % 14N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
14N2
CAS Number:
Molecular Weight:
28.01
UNSPSC Code:
12141903
PubChem Substance ID:

description

15N depleted

isotopic purity

99.99 atom % 14N

form

gas

mass shift

depleted

SMILES string

N#[14N]

InChI

1S/N2/c1-2/i1+0

InChI key

IJGRMHOSHXDMSA-IGMARMGPSA-N

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah G Pati et al.
Rapid communications in mass spectrometry : RCM, 30(6), 684-690 (2016-02-13)
Oxygen isotope fractionation of molecular O2 is an important process for the study of aerobic metabolism, photosynthesis, and formation of reactive oxygen species. The latter is of particular interest for investigating the mechanism of enzyme-catalyzed reactions, such as the oxygenation
Kyla Cochrane et al.
Microbial genomics, 6(2) (2019-10-30)
Here, we report comprehensive transcriptomic profiles from Fusobacterium nucleatum under conditions that mimic the first stages of bacterial infection in a highly differentiated adenocarcinoma epithelial cell line. Our transcriptomic in vitro adenocarcinoma approach allows us to measure the expression dynamics
Yuanzheng Chen et al.
Scientific reports, 8(1), 10670-10670 (2018-07-15)
Various nitrogen species in nitrides are fascinating since they often appear with these nitride as superconductors, hard materials, and high-energy density. As a typical complex, though iron nitride has been intensively studied, nitrogen species in the iron-nitrogen (Fe-N) compounds only
Paul Brouwer et al.
Frontiers in plant science, 8, 442-442 (2017-04-15)
Sustainable agriculture demands reduced input of man-made nitrogen (N) fertilizer, yet N
Adam C Martiny et al.
Science advances, 5(8), eaax0341-eaax0341 (2019-09-07)
Surface ocean phosphate is commonly below the standard analytical detection limits, leading to an incomplete picture of the global variation and biogeochemical role of phosphate. A global compilation of phosphate measured using high-sensitivity methods revealed several previously unrecognized low-phosphate areas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service