Skip to Content
Merck
All Photos(1)

Key Documents

606472

Sigma-Aldrich

4-Hydroxybenzoic acid-13C7

99 atom % 13C, 99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO13C6H413CO2H
CAS Number:
Molecular Weight:
145.07
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

solid

mp

214-215 °C (lit.)

mass shift

M+7

SMILES string

O[13C](=O)[13c]1[13cH][13cH][13c](O)[13cH][13cH]1

InChI

1S/C7H6O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H,(H,9,10)/i1+1,2+1,3+1,4+1,5+1,6+1,7+1

InChI key

FJKROLUGYXJWQN-BNUYUSEDSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service