Skip to Content
Merck
All Photos(1)

Key Documents

604380

Sigma-Aldrich

Paraformaldehyde-13C

≥99 atom % 13C, 99% (CP)

Synonym(s):

Formaldehyde-13C polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(13CH2O)n
Molecular Weight:
31.02
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

≥99 atom % 13C

Quality Level

Assay

99% (CP)

form

solid

mp

170 °C (dec.) (lit.)

mass shift

M+1

SMILES string

[X]O[13CH2]O[13CH2]O[13CH2][X]

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Flam. Sol. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

André K Eckhardt et al.
Nature chemistry, 10(11), 1141-1147 (2018-09-12)
Carbohydrates (CH2O)n are the formal adducts of carbon (atoms) to water with a repeating unit that structurally resembles H-C̈-OH (hydroxymethylene). Although hydroxymethylene has been suggested as a building block for sugar formation, it is a reactive species that had escaped

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service