604186
Benzoyl chloride-(phenyl-13C6)
99 atom % 13C, 99% (CP)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
13C6H5COCl
CAS Number:
Molecular Weight:
146.52
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % 13C
Assay
99% (CP)
bp
198 °C (lit.)
mp
-1 °C (lit.)
density
1.262 g/mL at 25 °C
mass shift
M+6
storage temp.
−20°C
SMILES string
ClC(=O)[13c]1[13cH][13cH][13cH][13cH][13cH]1
InChI
1S/C7H5ClO/c8-7(9)6-4-2-1-3-5-6/h1-5H/i1+1,2+1,3+1,4+1,5+1,6+1
InChI key
PASDCCFISLVPSO-IDEBNGHGSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
161.6 °F - closed cup
Flash Point(C)
72 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jenny-Marie T Wong et al.
Journal of chromatography. A, 1446, 78-90 (2016-04-17)
Widely targeted metabolomic assays are useful because they provide quantitative data on large groups of related compounds. We report a high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) method that utilizes benzoyl chloride labeling for 70 neurologically relevant compounds, including catecholamines
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service