Skip to Content
Merck
All Photos(1)

Key Documents

603546

Sigma-Aldrich

Methanol-13C

99 atom % 13C, <1% 18O

Synonym(s):

Methyl-13C alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3OH
Molecular Weight:
33.03
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:

isotopic purity

99 atom % 13C

bp

65.4 °C

density

0.815 g/mL at 25 °C

mass shift

M+1

SMILES string

[13CH3]O

InChI

1S/CH4O/c1-2/h2H,1H3/i1+1

InChI key

OKKJLVBELUTLKV-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service