602965
Water-17O
40-44.9 atom % 17O, 99% (CP)
Synonym(s):
17O Labeled water
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H217O
CAS Number:
Molecular Weight:
19.02
MDL number:
UNSPSC Code:
50202301
PubChem Substance ID:
Recommended Products
isotopic purity
40-44.9 atom % 17O
Assay
99% (CP)
mol wt
18.42 by atom % calculation (at 40 atom % 17O)
bp
100 °C (lit.)
mp
0 °C (lit.)
density
1.044 g/mL at 25 °C
bulk density
1.01
mass shift
M+1
SMILES string
[17OH2]
InChI
1S/H2O/h1H2/i1+1
InChI key
XLYOFNOQVPJJNP-OUBTZVSYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
(Normalized with respect to hydrogen)
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service