Skip to Content
Merck
All Photos(1)

Key Documents

600016

Sigma-Aldrich

3-Methoxy-13C,d3-benzyl-α-13C,α,α-d2 bromide

98 atom % D, 99 atom % 13C

Synonym(s):

1-Bromoethyl-3-methoxy-13C,d3-benzene-α-13C,α,α-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C6D5H4BrO
CAS Number:
Molecular Weight:
208.08
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % D

bp

152 °C (lit.)

density

1.486 g/mL at 25 °C

mass shift

M+7

SMILES string

[2H][13C]([2H])([2H])Oc1cccc(c1)[13C]([2H])([2H])Br

InChI

1S/C8H9BrO/c1-10-8-4-2-3-7(5-8)6-9/h2-5H,6H2,1H3/i1+1D3,6+1D2

InChI key

ZKSOJQDNSNJIQW-KJVVDLQLSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service