All Photos(1)
About This Item
Linear Formula:
DOCD2(CD2)2CD2OD
CAS Number:
Molecular Weight:
100.18
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
Recommended Products
isotopic purity
98 atom % D
mol wt
99.98 by atom % calculation
bp
230 °C (lit.)
density
1.128 g/mL at 25 °C (est.)(lit.)
mass shift
M+10
SMILES string
[2H]OC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])O[2H]
InChI
1S/C4H10O2/c5-3-1-2-4-6/h5-6H,1-4H2/i1D2,2D2,3D2,4D2,5D,6D
InChI key
WERYXYBDKMZEQL-MBXGXEIXSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service