Skip to Content
Merck
All Photos(1)

Key Documents

593532

Sigma-Aldrich

Fmoc-Leu-OH-13C6,15N

98 atom % 15N, 98 atom % 13C, 99% (CP)

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-L-Leucine-13C6,15N, L-Leucine-13C6,15N, N-Fmoc, 13C and 15N Labeled Fmoc-Leu

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(13CH3)213CH13CH213CH(15NH-Fmoc)13CO2H
CAS Number:
Molecular Weight:
360.36
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 13C
98 atom % 15N

Quality Level

Assay

99% (CP)

form

solid

optical activity

[α]20/D -25°, c = 0.5 in DMF

mp

152-156 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

mass shift

M+7

SMILES string

[13CH3][13CH]([13CH3])[13CH2][13C@H]([15NH]C(=O)OCC1c2ccccc2-c3ccccc13)[13C](O)=O

InChI

1S/C21H23NO4/c1-13(2)11-19(20(23)24)22-21(25)26-12-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h3-10,13,18-19H,11-12H2,1-2H3,(H,22,25)(H,23,24)/t19-/m0/s1/i1+1,2+1,11+1,13+1,19+1,20+1,22+1

InChI key

CBPJQFCAFFNICX-HNIVWWTASA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ksenia G Kuznetsova et al.
Journal of proteome research, 17(11), 3889-3903 (2018-10-10)
Adenosine-to-inosine RNA editing is one of the most common types of RNA editing, a posttranscriptional modification made by special enzymes. We present a proteomic study on this phenomenon for Drosophila melanogaster. Three proteome data sets were used in the study:
Arthur T Kopylov et al.
Journal of proteome research, 18(1), 120-129 (2018-11-28)
This work continues the series of the quantitative measurements of the proteins encoded by different chromosomes in the blood plasma of a healthy person. Selected Reaction Monitoring with Stable Isotope-labeled peptide Standards (SRM SIS) and a gene-centric approach, which is
Q Chen et al.
Journal of dairy science, 99(12), 9483-9492 (2016-10-25)
Fraud in milk and dairy products occurs when cow milk is added to sheep and goat milk for economic reasons. No reliable, selective, and sensitive method exists for quantifying the milk percentage of different species. This work reports the development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service