Skip to Content
Merck
All Photos(1)

Key Documents

592889

Sigma-Aldrich

Melamine-(triamine-15N3)

>80 atom % 15N (triamine), <20 atom % 15N (triazine), ≥97% (CP)

Synonym(s):

1,3,5-Triazine-2,4,6-triamine-15N3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H615N3N3
CAS Number:
Molecular Weight:
129.10
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

>80 atom % 15N (triamine)
<20 atom % 15N (triazine)

Assay

≥97% (CP)

mol wt

129.10 by atom % calculation (at 85 atom % 15N3 triamine, 15 atom % 15N3 triazine)

technique(s)

mass spectrometry (MS): suitable

mp

>300 °C (lit.)

mass shift

M+3

SMILES string

[15NH2]c1nc([15NH2])nc([15NH2])n1

InChI

1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)/i4+1,5+1,6+1

InChI key

JDSHMPZPIAZGSV-VMGGCIAMSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Urinary tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

572.0 °F - closed cup

Flash Point(C)

300 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tian Tian et al.
Molecular cell, 81(1), 198-211 (2020-12-10)
Replication fork reversal is a global response to replication stress in mammalian cells, but precisely how it occurs remains poorly understood. Here, we show that, upon replication stress, DNA topoisomerase IIalpha (TOP2A) is recruited to stalled forks in a manner

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service