Skip to Content
Merck
All Photos(2)

Key Documents

592587

Sigma-Aldrich

N-(2,4-Dimethylphenyl)formamide

97%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
SGD 226.00
500 MG
SGD 803.00

SGD 226.00


Availability

Available to ship onDecember 24, 2024Details


Request a Bulk Order

Select a Size

Change View
100 MG
SGD 226.00
500 MG
SGD 803.00

About This Item

Linear Formula:
(CH3)2C6H3NHCHO
CAS Number:
Molecular Weight:
149.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

SGD 226.00


Availability

Available to ship onDecember 24, 2024Details


Request a Bulk Order

Quality Level

Assay

97%

mp

114-118 °C (lit.)

functional group

amide

SMILES string

[H]C(=O)Nc1ccc(C)cc1C

InChI

1S/C9H11NO/c1-7-3-4-9(10-6-11)8(2)5-7/h3-6H,1-2H3,(H,10,11)

InChI key

JOFDPSBOUCXJCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service