Skip to Content
Merck
All Photos(1)

Key Documents

591742

Sigma-Aldrich

Phenol-2,4,6-d3,OD

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D3C6H2OD
CAS Number:
Molecular Weight:
98.14
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Quality Level

form

solid

bp

182 °C (lit.)

mp

40-42 °C (lit.)

density

1.116 g/mL at 25 °C

mass shift

M+4

SMILES string

[2H]Oc1c([2H])cc([2H])cc1[2H]

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H/i1D,4D,5D/hD

InChI key

ISWSIDIOOBJBQZ-FYPISFBKSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service