Skip to Content
Merck
All Photos(1)

Key Documents

590959

Sigma-Aldrich

Vinyl-13C2 Bromide

gas, 99 atom % 13C

Synonym(s):

Bromoethylene-13C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H213C13CHBr
CAS Number:
Molecular Weight:
108.93
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

form

gas

bp

16 °C/750 mmHg (lit.)

mp

-139 °C (lit.)

mass shift

M+2

SMILES string

Br[13CH]=[13CH2]

InChI

1S/C2H3Br/c1-2-3/h2H,1H2/i1+1,2+1

InChI key

INLLPKCGLOXCIV-ZDOIIHCHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service