Skip to Content
Merck
All Photos(1)

Key Documents

586900

Sigma-Aldrich

Ethylene-d3

98 atom % D, 99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D2C=CHD
CAS Number:
Molecular Weight:
31.07
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

99% (CP)

bp

-104 °C (lit.)

mass shift

M+3

SMILES string

[H]\C([2H])=C(\[2H])[2H]

InChI

1S/C2H4/c1-2/h1-2H2/i1D,2D2

InChI key

VGGSQFUCUMXWEO-APAIHEESSA-N

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-148.0 °F - closed cup

Flash Point(C)

-100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service