Skip to Content
Merck
All Photos(1)

Key Documents

586072

Sigma-Aldrich

Dodecylamine-15N

98 atom % 15N

Synonym(s):

Laurylamine-15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)1115NH2
CAS Number:
Molecular Weight:
186.34
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % 15N

Quality Level

form

solid

bp

247-249 °C

mp

27-29 °C (lit.)

density

0.810 g/mL

mass shift

M+1

SMILES string

CCCCCCCCCCCC[15NH2]

InChI

1S/C12H27N/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-13H2,1H3/i13+1

InChI key

JRBPAEWTRLWTQC-KCKQSJSWSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Oral - STOT SE 3

Target Organs

Gastrointestinal tract,Liver,Immune system, Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service