Skip to Content
Merck
All Photos(2)

Key Documents

576352

Sigma-Aldrich

Indium tin oxide coated glass slide, rectangular

surface resistivity 70-100 Ω/sq, slide

Synonym(s):

ITO, ITO coated slide, rectangular

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In2O3 · (SnO2)x
CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

surface resistivity

70-100 Ω/sq

L × W × thickness

75 mm × 25 mm × 1.1 mm

transmittance

>87%

refractive index

n20/D 1.517 (lit.)

SMILES string

O=[Sn]=O.O=[In]O[In]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium tin oxide (ITO) is widely used as transparent conducting oxides. Its major characteristics are electrical conductivity, optical transparency and the ease to be deposited as films.

Application

ITO cover slides may be used for closed digital microfluidic devices and as a transparent electrodes in display and other optoelectronic devices. Some other potential uses that have been reported are:
  • Eumelanin samples prepared on ITO covered glass substrates.
  • Interface windows of the liquid cell for optical beam deflection (OBD) sensing contain a 20-nm-thick indium tin oxide film.
  • Mesoporous TiO2 films were formed on ITO conductive glass substrates.

Physical properties

Thickness of ITO coating is 150-300 Å.
Resistance may increase to as high as 375 Ω when exposed to temperatures of 300 °C for 30 minutes or more.

To avoid damaging the thin film coating when determining which side is coated, lightly touch the barrelsof the probes to the same surface of a single-side coated product. Testing the alternate side will quicklyshow one with a low resistance as compared to the opposing side, which will exhibit an essentially infiniteresistance.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Online coupling of digital microfluidic devices with mass spectrometry detection using an eductor with electrospray ionization.
Baker CA and Roper MG
Analytical Chemistry, 2955-2960 (2012)
Preparation of non-annealed anatase TiO< sub> 2</sub> film on ITO substrate by anodizing in hot phosphate/glycerol electrolyte for dye-sensitized solar cells.
Tsuji E, et al.
Materials Letters, 91, 39-41 (2013)
High-resolution frequency-modulation atomic force microscopy in liquids using electrostatic excitation method.
Umeda K, et al.
Applied Physics Express, 3(6), 065205-065205 (2010)
Optical Fiber Refractometers based on Indium Tin Oxide Coatings with Response in the Visible Spectral Region
Zamarreno CR, et al.
Procedia Engineering, 25, 499-502 (2011)
Andreas Herrmann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(1), 117-124 (2008-12-17)
Application of an electric field to liquid crystalline film forming imines with negative dielectric anisotropy, such as N-(4-methoxybenzylidene)-4-butylaniline (MBBA, 1), results in the expulsion of compounds that do not participate in the formation of the liquid crystalline phase. Furthermore, amines

Articles

In this article, we demonstrate that bis-styrylbenzene derivatives show promising characteristics for very low lasing thresholds and discuss the design considerations for organic lasing molecules.

Organic photovoltaics (OPVs) represent a low-cost, lightweight, and scalable alternative to conventional solar cells. While significant progress has been made in the development of conventional bulk heterojunction cells, new approaches are required to achieve the performance and stability necessary to enable commercially successful OPVs.

A transparent conductive electrode (TCE) is an essential component of various optoelectronic devices such as solar cells, liquid-crystal displays (LCD), light-emitting diodes (LED), and touch screens.

A transparent conductive electrode (TCE) is an essential component of various optoelectronic devices such as solar cells, liquid-crystal displays (LCD), light-emitting diodes (LED), and touch screens.

See All

Protocols

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service