Skip to Content
Merck
All Photos(2)

Key Documents

571555

Sigma-Aldrich

Potassium p-tolyltrifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4BF3K
CAS Number:
Molecular Weight:
198.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

SMILES string

[K+].Cc1ccc(cc1)[B-](F)(F)F

InChI

1S/C7H7BF3.K/c1-6-2-4-7(5-3-6)8(9,10)11;/h2-5H,1H3;/q-1;+1

InChI key

KRWDYXJWQBTBAH-UHFFFAOYSA-N

Application

Potassium p-tolyltrifluoroborate can be used:
  • As a precursor/starting material for the synthesis of biaryl compounds by reacting with various aryl halides using Pd/C catalyst.
  • As a reagent in the carbonylative arylation of vinyl ketones via 1,4-addition.
  • As a substrate in the synthesis of primary arylamines by reacting with hydroxylamine-O-sulfonic acid under metal-free conditions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbonylative 1, 4-addition of potassium aryltrifluoroborates to vinyl ketones
Sauthier M, et al.
New. J. Chem., 33(5), 969-971 (2009)
Palladium on carbon-catalyzed cross-coupling of aryl halides with potassium p-tolyltrifluoroborate in air
LeBlond CR, et al.
Synthetic Communications, 39(4), 636-640 (2009)
Oxygen-promoted Pd/C-catalyzed Suzuki-Miyaura reaction of potassium aryltrifluoroborates
Liu C, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 27(5), 631-634 (2016)
Sonication and Microwave-Assisted Primary Amination of Potassium Aryltrifluoroborates and Phenylboronic Acids under Metal-Free Conditions
Kuik D, et al.
Synthesis, 49(11), 2555-2561 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service