Skip to Content
Merck
All Photos(2)

Key Documents

570311

Sigma-Aldrich

Poly(ethylene glycol)-block-poly(ε−caprolactone) methyl ether

PCL average Mn ~13,000, PEG average Mn ~5,000

Synonym(s):

Poly(ethylene glycol) methyl ether-block-poly(ε−caprolactone)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

Quality Level

mol wt

PCL average Mn ~13,000
PEG average Mn ~5,000
average Mn ~18,000 (total)

degradation timeframe

>12 months

mp

54.2-55.9 °C (lit.)
58.8-60.7 °C

Mw/Mn

<1.4

PDI

≤1.4

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Biocompatible diblock copolymer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service