570079
2-Isopropyl-5-methylcyclohexanol-1,2,6,6-d4
98 atom % D
Synonym(s):
Menthol-plus its 3 isomers-1,2,6,6-d4
About This Item
Recommended Products
isotopic purity
98 atom % D
Assay
99% (CP)
form
solid
mass shift
M+4
SMILES string
OC1([2H])C(C(C)C)([2H])CCC(C)C1([2H])[2H]
InChI
1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/i6D2,9D,10D
InChI key
NOOLISFMXDJSKH-JKQVIGAJSA-N
Related Categories
Application
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service