Skip to Content
Merck
All Photos(1)

Key Documents

558753

Sigma-Aldrich

1,4-Diacetoxy-2-bromobenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H3CCO2)2C6H3Br
CAS Number:
Molecular Weight:
273.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

70-75 °C (lit.)

functional group

bromo
ester

SMILES string

CC(=O)Oc1ccc(OC(C)=O)c(Br)c1

InChI

1S/C10H9BrO4/c1-6(12)14-8-3-4-10(9(11)5-8)15-7(2)13/h3-5H,1-2H3

InChI key

XRIFNWOHWJOCQG-UHFFFAOYSA-N

General description

1,4-Diacetoxy-2-bromobenzene can be prepared by reacting 1,4-benzoquinone with zinc bromide in the presence of acetic anhydride.

Application

1,4-Diacetoxy-2-bromobenzene may be used to synthesize bromohydroquinone and (Sp,S)-1-(2,5-diacetoxyphenyl)-2-(p-tolylsulfinyl)ferrocene.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, Synthesis, and Evaluation of Synthetic Mimics of Cell Surface Receptors, 212-212 (2007)
Total synthesis of siccayne.
Pinault M, et al.
Synthesis, 10, 935-937 (1990)
Highly Selective Negishi Cross-Coupling Reaction of a Zinc-Metallated Ferrocenyl p-Tolyl Sulfoxide: New Chiral Ferrocene-Based Quinone Ligands.
Cotton HK, et al.
European Journal of Organic Chemistry, 15, 2756-2763 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service