All Photos(1)
About This Item
Linear Formula:
CH3OC6H4CH2NCO
CAS Number:
Molecular Weight:
163.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.5320 (lit.)
density
1.136 g/mL at 25 °C (lit.)
functional group
amine
isocyanate
storage temp.
2-8°C
SMILES string
COc1cccc(CN=C=O)c1
InChI
1S/C9H9NO2/c1-12-9-4-2-3-8(5-9)6-10-7-11/h2-5H,6H2,1H3
InChI key
QWGAJMDRYUIPIT-UHFFFAOYSA-N
Related Categories
General description
3-Methoxybenzyl isocyanate (3MBITC) is the degradation product of glucolimnanthin, a glucosinolate found in meadowfoam (Limnanthes alba) seed oil.
Application
3-Methoxybenzyl isocyanate may be used to synthesize the following thiourea derivatives:
- 1-(3-methoxybenzyl)-3-dodecyl thiourea
- 1-(3-methoxybenzyl)-3-octyl thiourea
- 1,3-di(3-methoxybenzyl) thiourea
- 1-(3-methoxybenzyl)-3-octadecyl thiourea
Caution
Refrigerate
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
80.1 °F - closed cup
Flash Point(C)
26.7 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
1, 3-di (3-Methoxybenzyl) thiourea and related lipid antioxidants.
Abbott TP, et al.
Industrial Crops and Products, 16(1), 43-57 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service