All Photos(1)
About This Item
Linear Formula:
HOCH2CH2NHC(O)NH2
CAS Number:
Molecular Weight:
104.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
solid
mp
164-169 °C (lit.)
functional group
amine
hydroxyl
SMILES string
NC(=O)NCCO
InChI
1S/C3H8N2O2/c4-3(7)5-1-2-6/h6H,1-2H2,(H3,4,5,7)
InChI key
CLAHOZSYMRNIPY-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ala'a F Eftaiha et al.
Physical chemistry chemical physics : PCCP, 22(21), 11829-11837 (2020-05-20)
(Thio)Urea scaffolds are best known for their importance as intermediates in organic synthesis. In this work, a mechanistic study of the reaction between urea (U), (2-hydroxyethyl)urea (U-EtOH) and thiourea (tU)/NaH in DMSO with CO2 was carried out. While both U/tU
Fan C Wang et al.
Journal of cosmetic science, 68(2), 159-172 (2018-04-06)
A petroleum-free skin cream was developed using food-grade ingredients. The rheological and sensorial properties of this petroleum-free skin cream were compared to a commercially available petroleum-based skin cream. Specifically, large-amplitude oscillatory shear (LAOS) characterization of the two skin creams was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service