548227
2-Chloro-4-pyridinecarbonitrile
97%
Synonym(s):
2-Chloro-4-cyanopyridine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
mp
69-73 °C (lit.)
functional group
chloro
nitrile
SMILES string
Clc1cc(ccn1)C#N
InChI
1S/C6H3ClN2/c7-6-3-5(4-8)1-2-9-6/h1-3H
InChI key
QRXBTPFMCTXCRD-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service