Skip to Content
Merck
All Photos(1)

Key Documents

54340

Sigma-Aldrich

2-Hydroxyethylhydrazine

≥95% (GC)

Synonym(s):

2-Hydrazinoethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2NHNH2
CAS Number:
Molecular Weight:
76.10
Beilstein:
1731669
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (GC)

refractive index

n20/D 1.493 (lit.)
n20/D 1.493

bp

155-160 °C/32 mmHg (lit.)

mp

−70 °C (lit.)

density

1.123 g/mL at 25 °C (lit.)

functional group

amine
hydrazine
hydroxyl

SMILES string

NNCCO

InChI

1S/C2H8N2O/c3-4-1-2-5/h4-5H,1-3H2

InChI key

GBHCABUWWQUMAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hydroxyethylhydrazine reacts with β-diketones having strong electron-withdrawing substituents to yield 5-hydroxy-4,5-dihydropyrazoles.

Other Notes

Sales restrictions may apply

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Reaction of 2-hydroxyethylhydrazine with a trifluoromethyl-?-diketone: Study and structural characterization of a new 5-hydroxy-5-trifluoromethyl-4, 5-dihydropyrazole intermediate.
Montoya V, et al.
Journal of Fluorine Chemistry, 128(9), 1007-1011 (2007)
Smaail Radi et al.
Molecules (Basel, Switzerland), 21(8), doi:10-doi:10 (2016-08-17)
A pyridylpyrazole bearing a hydroxyethyl substituent group has been synthesized by condensation of (Z)-4-hydroxy-4-(pyridin-2-yl)but-3-en-2-one with 2-hydroxyethylhydrazine. The compound was well characterized and its structure confirmed by single crystal X-ray diffraction. Density functional calculations have been performed using DFT method with
A new method for the synthesis of 1, 4, 5-oxadiazocines and its application in the structure modification of natural products.
Liu W, et al.
Tetrahedron Letters, 46(46), 8009-8012 (2005)
C Andriamampandry et al.
Journal of neurochemistry, 56(6), 1845-1850 (1991-06-01)
The sequential methylation of ethanolamine and its phosphorylated derivatives has been studied with chick neurons in culture in the presence of several pharmacological agents. Incubation with [3H]ethanolamine in the presence of monomethylethanolamine and dimethylethanolamine indicated that in these neurons the
Gregory M Sandala et al.
Journal of the American Chemical Society, 127(24), 8856-8864 (2005-06-16)
Ab initio molecular orbital calculations have been used to study the mechanism of suicide inactivation of ethanolamine ammonia-lyase induced by three different substrate analogues. Analysis of the normal catalytic mechanism with 2-aminoethanol (ethanolamine) as substrate predicts that both the hydrogen-abstraction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service